4-(2-Pyridylazo)resorcinol 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(2'-Pyridylazo)resorcinol

41859-0010 41859-0050 41859-0250
TS41859-0010EA 35 USD
TS41859-0010 TS41859-0050 TS41859-0250
4-(2-Pyridylazo)resorcinol 97+%
4-(2-Pyridylazo)resorcinol
Formula: C₁₁H₉N₃O₂
MW: 215.21
Melting Pt: 188 °C
Storage Temperature: Ambient
MDL Number: MFCD00044237
CAS Number: 1141-59-9
EINECS: 214-528-6

Order Now

Specification Test Results

Appearance Orange Powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR