4-Phenylazophenol 97%

Supplier: Thermo Scientific Chemicals
Warning

41736-0250 41736-1000
TS41736-0250EA 54 USD
TS41736-0250 TS41736-1000
4-Phenylazophenol 97%
4-Phenylazophenol
Formula: C₁₂H₁₀N₂O
Melting Pt: 150…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00002330
CAS Number: 1689-82-3
EINECS: 216-880-6

Order Now

Specification Test Results

Appearance Brown Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR