Phenolphthalein disulfate tripotassium salt trihydrate 98%, pure

Supplier: Thermo Scientific Chemicals

41722-0050 41722-0100
TS41722-0050EA 198 USD
TS41722-0050 TS41722-0100
Phenolphthalein disulfate tripotassium salt trihydrate 98%, pure
Phenolphthalein disulfate tripotassium salt trihydrate
Formula: C₂₀H₁₃K₃O₁₁S₂.3H₂O
Melting Pt: 230 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00150195
CAS Number: 1080664-81-8

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR