Pararosaniline hydrochloride 98%, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
41680-0050
41680-0250
41680-1000
TS41680-0050EA
29
USD
TS41680-0050
TS41680-0250
TS41680-1000
Pararosaniline hydrochloride 98%, pure
Pararosaniline hydrochloride
Formula:
C₁₉H₁₈ClN₃ Melting Pt: 268…270 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00001657 CAS Number: 569-61-9 EINECS: 209-321-2 |
Specification Test Results
Appearance | Green to dark green Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤10 % (110°C, 1 h) |
HPLC | ≥97.5 % |
UV Lambda max | 543 to 549 nm (c=0.0003%, 50% EtOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...