Nitro blue tetrazolium chloride (NBT) 90%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5,5'-Diphenyl-3,3'-bis(4-nitrophenyl)-2,2'-(3,3'-dimethoxybiphenyl-4,4'-ylene)ditetrazolium dichloride

41592-0010 41592-5000
TS41592-0010EA 174 USD
TS41592-0010 TS41592-5000
Nitro blue tetrazolium chloride (NBT) 90%
Nitro blue tetrazolium chloride (NBT)
Formula: C₄₀H₃₀Cl₂N₁₀O₆
MW: 817.65 g/mol
Melting Pt: 189 °C
Storage Temperature: Freezer
MDL Number: MFCD00012159
CAS Number: 298-83-9
EINECS: 206-067-4

Order Now

Specification Test Results

Appearance Yellow Crystalline powder or crystals
Infrared spectrum Conforms
Titration Argentometric ≥88 %
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR