Nile blue A, pure, certified
Supplier: Thermo Scientific Chemicals
Synonyms:
Nile blue A sulphate, Nile blue sulphate, 5-Amino-9-(diethylamino)benzo[a]phenoxazin-7-ium sulphate, Basic Blue 12
41569-0100
41569-0500
TS41569-0100EA
53
USD
TS41569-0100
TS41569-0500
Nile blue A, pure, certified
Nile blue A
Formula:
C₄₀H₄₀N₆O₆S MW: 732.84 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00064529 CAS Number: 3625-57-8 |
Specification Test Results
Appearance | Dark green to black Powder |
Infrared spectrum | Conforms |
Dye content | ≥70 % |
UV Lambda max | 635 to 646 nm |
Absorption Ratio P-15/P+15 | 1.01 to 1.16 |
BSC Certification | Passes |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...