Methyl orange ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium salt, Methyl orange sodium salt, Helianthine

41466-0100 41466-0250 41466-1000
TS41466-0100EA 27 USD
TS41466-0100 TS41466-0250 TS41466-1000
Methyl orange ACS
Methyl orange
Formula: C₁₄H₁₄N₃NaO₃S
MW: 327.34 g/mol
Boiling Pt: 90
Melting Pt: >300 °C
Flash Pt: 38
Storage Temperature: Ambient
MDL Number: MFCD00007502
CAS Number: 547-58-0
EINECS: 208-925-3
UN: 3143
ADR: 6.1 III
Merck Index: 13,06127

Order Now

Specification Test Results

Appearance Orange Crystalline powder
Infrared spectrum Conforms
Visual transition interval From pH 3.2 (pink or red) to pH 4.4 (yellow)
Clarity of solution Passes test
Stabilizer 100 to 400 ppm MEHQ


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR