Methyl green zinc chloride salt, pure, certified

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: CI 42590, Ethyl Green, Methyl green zinc chloride double salt, Methyl Green

41438-0250 41438-1000
TS41438-0250EA 109 USD
TS41438-0250 TS41438-1000
Methyl green zinc chloride salt, pure, certified
Methyl Green zinc chloride salt
Formula: C₂₇H₃₅BrClN₃·xZnCl₂
MW: 653.24
Melting Pt: 233 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00151094
CAS Number: 7114-03-6
EINECS: 230-415-4

Order Now

Specification Test Results

Appearance Dark green to dark brown-red Powder
Infrared spectrum Conforms
Dye content ≥85 %
UV Lambda max 630 to 635 nm (in water)
Absorption Ratio P-15/P+15 0.86 to 1.11
Visual transition interval from pH 0.2 (yellow) to pH 1.8 (blue)
BSC Certification Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR