Magnesium bromide, anhydrous 98%, pure

Supplier: Thermo Scientific Chemicals
Warning

41339-0010 41339-0050
TS41339-0010EA 24 USD
TS41339-0010 TS41339-0050
Magnesium bromide, anhydrous 98%, pure
Magnesium bromide
Formula: Br₂Mg
MW: 184.11
Melting Pt: 700
Density: 3.72
Storage Temperature: Ambient
MDL Number: MFCD00011105
CAS Number: 7789-48-2
Merck Index: 12,05695

Order Now

Specification Test Results

Appearance White to grey Powder or chunks
Titration Argentometric 97.5 to 102.5 %
Refractive index 1.5680 to 1.5700 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR