Sulforhodamine B acid chloride, pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Lissamine rhodamine B sulphonyl chloride

41323-0010 41323-0050
TS41323-0010EA 415 USD
TS41323-0010 TS41323-0050
Sulforhodamine B acid chloride, pure
Sulforhodamine B acid chloride
Formula: C₂₇H₂₉ClN₂O₆S₂
Storage Temperature: Refrigerator
CAS Number: 62796-29-6

Order Now

Specification Test Results

Appearance Dark green Crystalline powder
Infrared spectrum Conforms
UV Lambda max 561 to 567 nm (in CH2Cl2)
Molar extinction coefficient ≥85000 l/(mol.cm) (lambda max)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR