Indocyanine green, pure laser grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
IR-125, 4,5-benzoindotricarbocyanine, IC Green, Cardiogreen, Sodium 4-(2-(7-(1,1-dimethyl-3-(4-sulphonatobutyl)-1H-benzo[e]indol-2(3H)-ylidene)hepta-1,3,5-trien-1-yl)-1,1-dimethyl-1H-benzo[e]indol-3-ium-3-yl)butane-1-sulphonate, Foxgreen
41254-0020
41254-1000
41254-5000
TS41254-0020EA
2033
USD
TS41254-0020
TS41254-1000
TS41254-5000
Indocyanine green, pure laser grade
Indocyanine green
Formula:
C₄₃H₄₇N₂NaO₆S₂ MW: 774.99 g/mol Melting Pt: 235 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00013078 CAS Number: 3599-32-4 |
Specification Test Results
Appearance | Green to brown Crystalline powder or crystals |
Infrared spectrum | Conforms |
UV Lambda max | 784 nm (in Methanol) |
Molar absorption coefficient | ≥230000 l/(mol.cm) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...