Pyranine 98%

Supplier: Thermo Scientific Chemicals
Warning

41218-0010 41218-0050 41218-0250
TS41218-0010EA 116 USD
TS41218-0010 TS41218-0050 TS41218-0250
Pyranine 98%
Pyranine
Formula: C₁₆H₇Na₃O₁₀S₃
MW: 524.371 g/mol
Density: 2.15 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00037575
CAS Number: 6358-69-6

Order Now

Specification Test Results

Appearance Yellow to yellow-green Powder or crystals
Infrared spectrum Conforms
UV spectrophotometry ≥97.5 %
UV Lambda max 450 to 458 nm (in 0.01N NaOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR