Hydrazine dihydrochloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hydrazine Dihydrochloride, Hydrazinium dichloride

41176-0025 41176-1000 41176-5000
TS41176-0025EA 682 USD
TS41176-0025 TS41176-1000 TS41176-5000
Hydrazine dihydrochloride 99%
Hydrazine dihydrochloride
Formula: H₆Cl₂N₂
MW: 104.97 g/mol
Melting Pt: 189
Storage Temperature: Ambient
MDL Number: MFCD00064543
CAS Number: 5341-61-7
EINECS: 226-283-2
UN: 3288
ADR: 6.1 III
Merck Index: 13,04789

Order Now

Specification Test Results

Appearance White Powder and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR