Haematoxylin 85%, pure, certified

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: DcH-48, Hydroxybrazilin, (6aS,11bR)-6,6a,7,11b-Tetrahydroindeno[2,1-c]chromene-3,4,6a,9,10-pentaol, NSC 2785, Natural Black I, Hydroxybrasilin, (+)-Hematoxylin

41116-0250 41116-1000 41116-5000
TS41116-0250EA 116 USD
TS41116-0250 TS41116-1000 TS41116-5000
Haematoxylin 85%, pure, certified
Hematoxylin
Formula: C₁₆H₁₄O₆
MW: 302.28 g/mol
Melting Pt: 100…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00078111
CAS Number: 517-28-2
EINECS: 208-237-3
Merck Index: 13,04654

Order Now

Specification Test Results

Appearance Light beige to brown Powder
Infrared spectrum Conforms
Loss on drying ≤10 % (1 g, 105°C, 3 h)
UV spectrophotometry ≥80 % (On dry basis)
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR