Fluorescein disodium salt dihydrate 90%, pure

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(6-hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid disodium salt dihydrate, Uranine dihydrate, C.I. 45350 dihydrate, NaFluo dihydrate, Acid Yellow 73 dihydrate, Disodium 2-(3-oxo-6-oxidoxanthen-9-yl)benzoate dihydrate, Sodium fluorescein dihydrate, Yellow No. 8 dihydrate, NaFl dihydrate

41064-1000 41064-5000
TS41064-1000EA 33 USD
TS41064-1000 TS41064-5000
Fluorescein disodium salt dihydrate 90%, pure
Fluorescein disodium salt dihydrate
Formula: C₂₀H₁₀Na₂O₅·2H₂O
MW: 412.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150814
CAS Number: 1264198-62-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=949f2473a2f0c6f8d00fd80ca2f80b730bc8076a&vid=fe174603ab8c1727364c1e09d665a1ad5a6d9106&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Orange-red to green-brown to dark brown Powder
Infrared spectrum Conforms
Water 6.5 to 10 % (K.F.)
UV spectrophotometry ≥88 % (on dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR