Calcein

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Oftasceina, Fluorescein Complexon, Oftasceinum, 2,7-Bis[bis(carboxymethyl)aminomethyl]fluorescein, Oftasceine, 2,2',2'',2'''-(((3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-2',7'-diyl)bis(methylene))bis(azanetriyl))tetraacetic acid, Fluorexon, Bis[N,N-bis(carboxymethyl)aminomethyl]fluorescein, NSC 298193

41063-0050 41063-0250
TS41063-0050EA 83 USD
TS41063-0050 TS41063-0250
Calcein
Calcein
Formula: C₃₀H₂₆N₂O₁₃
MW: 622.53 g/mol
Melting Pt: 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00005049
CAS Number: 1461-15-0
EINECS: 215-957-1

Order Now

Specification Test Results

Appearance Yellow to orange-brown Powder
Infrared spectrum Conforms
UV spectrum Complies
Suitability (for Ca determination) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR