Fast green FCF, pure, certified

Supplier: Thermo Scientific Chemicals
Warning

41053-0250 41053-1000
TS41053-0250EA 173 USD
TS41053-0250 TS41053-1000
Fast green FCF, pure, certified
Fast green FCF
Formula: C₃₇H₃₄N₂Na₂O₁₀S₃
Melting Pt: 290 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013053
CAS Number: 2353-45-9
EINECS: 219-091-5
Merck Index: 13,03970

Order Now

Specification Test Results

Appearance Bordeaux to brown Powder
Infrared spectrum Conforms
Dye content ≥85 %
UV Lambda max 622 to 626 nm
Absorption Ratio P-15/P+15 0.98 to 1.20
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR