Methyldiphenylphosphine 99%

Supplier: Thermo Scientific Chemicals
Warning

40897-0050 40897-0250 40897-1000
TS40897-0050EA 77 USD
TS40897-0050 TS40897-0250 TS40897-1000
Methyldiphenylphosphine 99%
Methyldiphenylphosphine
Formula: C₁₃H₁₃P
Boiling Pt: 118…120 °C (2.2 mmHg)
Density: 1.076 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008508
CAS Number: 1486-28-8
EINECS: 216-065-5

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6230 to 1.6260 (20°C, 589 nm)
GC ≥98.5 %
Pore volume 0.70 to 0.85 ml/g
Carbon (C) ≥20.7 %
Volatile matter ≤4 % (110°C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR