Dimethyl glyoxime 99+% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Diacetyl glyoxime, Diacetyl dioxime, 2,3-Butanedione dioxime

40833-0025 40833-0250 40833-1000 40833-5000
TS40833-0025EA 949 USD
TS40833-0025 TS40833-0250 TS40833-1000 TS40833-5000
Dimethyl glyoxime 99+% ACS
Dimethyl glyoxime
Formula: CH₃C(=NOH)C(=NOH)CH₃
MW: 116.12 g/mol
Boiling Pt: 53 °C (8 mmHg)
Melting Pt: 239…242 °C
Storage Temperature: Ambient
MDL Number: MFCD00002117
CAS Number: 95-45-4
EINECS: 202-420-1
UN: 2926
ADR: 4.1,III
Merck Index: 13,03272

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 239 to 242 °C (Dec)
Residue after ignition ≤0.05 %
Gravimetry ≥99.0 %
Suitability (for Ni determination) Passes test
Insoluble matter ≤0.05 % (in ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR