Dimethyldioctadecylammonium bromide (DDA) 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dimethyldioctadecylammonium Bromide

40826-0050 40826-0250 40826-1000
TS40826-0050EA 79 USD
TS40826-0050 TS40826-0250 TS40826-1000
Dimethyldioctadecylammonium bromide (DDA) 99+%
Dimethyldioctadecylammonium bromide (DDA)
Formula: C₃₈H₈₀BrN
Melting Pt: 151…157 °C
Density: 1.047 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00041975
CAS Number: 3700-67-2
EINECS: 223-037-6

Order Now

Specification Test Results

Appearance White to almost white Powder and/or chunks
Infrared spectrum Conforms
Melting point ≥150 °C clear point
Titration Argentometric 99.0 to 101.0 %
TLC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR