Didodecyldimethylammonium bromide 99%

Supplier: Thermo Scientific Chemicals
Warning

40712-0250 40712-1000
TS40712-0250EA 210 USD
TS40712-0250 TS40712-1000
Didodecyldimethylammonium bromide 99%
Didodecyldimethylammonium bromide
Formula: C₂₆H₅₆BrN
MW: 462.65 g/mol
Melting Pt: 161…164 °C
Storage Temperature: Ambient
MDL Number: MFCD00041969
CAS Number: 3282-73-3
EINECS: 221-923-7

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals
Infrared spectrum Conforms
Melting point 158 to 163 °C
Titration Argentometric ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR