Decyltrimethylammonium bromide 99%

Supplier: Thermo Scientific Chemicals
Warning

40623-0250 40623-1000
TS40623-0250EA 107 USD
TS40623-0250 TS40623-1000
Decyltrimethylammonium bromide 99%
Decyltrimethylammonium bromide
Formula: C₁₃H₃₀BrN
Melting Pt: 243 °C
Storage Temperature: Ambient
MDL Number: MFCD00041973
CAS Number: 2082-84-0
EINECS: 218-219-7

Order Now

Specification Test Results

Appearance White to beige Powder and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR