Cyclohexyldiphenylphosphine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diphenylcyclohexylphosphine

40610-0010 40610-0050
TS40610-0010EA 38 USD
TS40610-0010 TS40610-0050
Cyclohexyldiphenylphosphine 98%
Cyclohexyldiphenylphosphine
Formula: C₁₈H₂₁P
MW: 268.33 g/mol
Melting Pt: 53 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00046360
CAS Number: 6372-42-5
EINECS: 228-904-2

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR