o-Cresolphthalein complexone tetrasodium salt, pure
Supplier: Thermo Scientific Chemicals
Synonyms:
o-Cresolphthalein Complexone sodium salt
40575-0050
40575-0250
TS40575-0050EA
50
USD
TS40575-0050
TS40575-0250
o-Cresolphthalein complexone tetrasodium salt, pure
o-Cresolphthalein complexone tetrasodium salt
Formula:
C₃₂H₂₈N₂Na₄O₁₂ MW: 724.54 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00066371 CAS Number: 62698-54-8 |
Specification Test Results
Appearance | Purple Powder or crystals |
Infrared spectrum | Conforms |
Suitability | (for use as reagent for Ca) Passes test |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...