Coumarin 343, pure laser grade
Supplier: Thermo Scientific Chemicals
Synonyms:
2,3,6,7-Tetrahydro-11-oxo-1H,5H,11H-[1]benzopyrano[6,7,8-ij]quinolizine-10-carboxylic Acid, 11-oxo-2,3,5,6,7,11-Hexahydro-1H-pyrano[2,3-f]pyrido[3,2,1-ij]quinoline-10-carboxylic acid
40568-1000
40568-5000
TS40568-1000EA
53
USD
TS40568-1000
TS40568-5000
Coumarin 343, pure laser grade
Coumarin 343
Formula:
C₁₆H₁₅NO₄ Melting Pt: 240 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051335 CAS Number: 55804-65-4 |
Specification Test Results
Appearance | Orange Crystalline powder |
Infrared spectrum | Conforms |
UV Lambda max | 409 nm (in Basic Ethanol) |
Fluorescence max | 470 nm (in Basic Ethanol) |
Optimum lasing | 518 nm (in Methanol) |
Suitability | (for use as a laser dye) Passes test |
Molar absorption coefficient | 19900 l/(mol.cm) (at Lambda max) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...