Coumarin 314 99%, pure

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl 2,3,6,7-Tetrahydro-11-oxo-1H,5H,11H-[1]benzopyrano[6,7,8-ij]quinolizine-10-carboxylate, Coumarin 504, Ethyl 11-oxo-2,3,5,6,7,11-hexahydro-1H-pyrano[2,3-f]pyrido[3,2,1-ij]quinoline-10-carboxylate

40562-0025 40562-1000 40562-5000
TS40562-0025EA 506 USD
TS40562-0025 TS40562-1000 TS40562-5000
Coumarin 314 99%, pure
Coumarin 314
Formula: C₁₈H₁₉NO₄
Melting Pt: 140…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00051334
CAS Number: 55804-66-5

Order Now

Specification Test Results

Appearance Yellow to orange Crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %
UV Lambda max 435 to 439 nm (in Ethanol)
Molar absorption coefficient ≥45000 l/(mol.cm) (at Lambda max)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR