3-(2-Benzothiazolyl)-7-(diethylamino)coumarin 98%, pure

Supplier: Thermo Scientific Chemicals
Warning

40550-0025 40550-5000
TS40550-0025EA 142 USD
TS40550-0025 TS40550-5000
3-(2-Benzothiazolyl)-7-(diethylamino)coumarin 98%, pure
3-(2-Benzothiazolyl)-7-(diethylamino)coumarin
Formula: C₂₀H₁₈N₂O₂S
MW: 350.4341 g/mol
Melting Pt: 205…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00041869
CAS Number: 38215-36-0
EINECS: 253-830-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Orange Powder
Infrared spectrum Conforms
UV spectrophotometry ≥97.5 %
UV Lambda max 455 to 461 nm (0.005g/l,Ethanol)
Molar absorption coefficient ≥50000 l/(mol.cm) (at Lambda max)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR