Bis(cyclopentadienyl)cobalt 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
40532-0010
40532-0100
TS40532-0010EA
72
USD
TS40532-0010
TS40532-0100
Bis(cyclopentadienyl)cobalt 98%
Bis(cyclopentadienyl)cobalt
Formula:
C₁₀H₁₀Co Melting Pt: 173 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00013749 CAS Number: 1277-43-6 EINECS: 215-061-0 UN: 1325 ADR: 4.1,II |
Specification Test Results
Appearance | Black Metallic crystals |
Infrared spectrum | Conforms |
Elemental Analysis | ≥97.5 % |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...