5(6)-Carboxyfluorescein

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5(6)-FAM, 3′,6′-Dihydroxy-3-oxospiro[isobenzofuran-1(3H),9′-[9H]xanthene]-ar-carboxylic acid

40410-0050 40410-5000
TS40410-0050EA 164 USD
TS40410-0050 TS40410-5000
5(6)-Carboxyfluorescein
5(6)-Carboxyfluorescein
Formula: C₄₂H₂₄O₁₄
MW: 752.63 g/mol
Melting Pt: 275 °C
Storage Temperature: Ambient
MDL Number: MFCD00151081
CAS Number: 72088-94-9

Order Now

Specification Test Results

Appearance Orange to brown Crystalline powder and/or chunks
Infrared spectrum Conforms
Molar absorption coefficient ≥72000 l/(mol.cm) (at Lamda max 492 nm, in 0.01N NaOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR