Brucine sulfate heptahydrate 98+% ACS

Supplier: Thermo Scientific Chemicals
Danger

40329-0100 40329-0250
TS40329-0100EA 38 USD
TS40329-0100 TS40329-0250
Brucine sulfate heptahydrate 98+% ACS
Brucine sulfate heptahydrate
Formula: C₂₃H₂₆N₂O₄
MW: 887.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013472
CAS Number: 60583-39-3
UN: 2811
ADR: 6.1 I

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤13.0 % (1 g, 105°C, 6 h)
Residue after ignition ≤0.1 %
Titration with HClO4 ≥98.0 % (on dry substance)
Sensitivity (to nitrate) Passes test
Clarity of solution Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR