2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-((5-Bromopyridin-2-yl)diazenyl)-5-(diethylamino)phenol, 2-(4-Diethylamino-2-hydroxyphenylazo)-5-bromopyridine

40322-0010 40322-0050
TS40322-0010EA 163 USD
TS40322-0010 TS40322-0050
2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol 98%
2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol
Formula: C₁₅H₁₇BrN₄O
MW: 349.23 g/mol
Melting Pt: 155…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00006255
CAS Number: 14337-53-2

Order Now

Specification Test Results

Appearance Red to dark red to purple red Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Solubility complete dissolution within max 5 min
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR