Bromophenol blue sodium salt ACS

Supplier: Thermo Scientific Chemicals

Synonyms: Bromophenol blue sodium, Sodium α-(3,5-dibromo-4-oxo-2,5-cyclohexadienylidene)-α-(3,5-dibromo-4-hydroxyphenyl)toluenesulphonate, Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)phenolate, Bromophenol blue water soluble, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2,6-dibromophenol] S,S-dioxide sodium salt

40316-0050 40316-0100 40316-0500
TS40316-0050EA 36 USD
TS40316-0050 TS40316-0100 TS40316-0500
Bromophenol blue sodium salt ACS
Bromophenol blue sodium salt
Formula: C₁₉H₉Br₄NaO₅S
MW: 691.95 g/mol
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013793
CAS Number: 34725-61-6
EINECS: 252-170-2

Order Now

Specification Test Results

Appearance Dark green to brown-black to blue-black Powder
Infrared spectrum Conforms
Visual transition interval from pH 3.0 (yellow) to pH 4.6 (blue)
Clarity of solution Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR