Bromocresol green sodium salt ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bromocresol green water soluble, Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-2,2-dioxido-3H-benzo[d][1,2]oxathiol-3-yl)-3-methylphenolate

40294-0050 40294-0100 40294-0500
TS40294-0050EA 56 USD
TS40294-0050 TS40294-0100 TS40294-0500
Bromocresol green sodium salt ACS
Bromocresol green sodium salt
Formula: C₂₁H₁₃Br₄NaO₅S
MW: 720 g/mol
Melting Pt: 230 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00148898
CAS Number: 62625-32-5
EINECS: 263-657-4

Order Now

Specification Test Results

Appearance Dark brown to green-black Powder
Infrared spectrum Conforms
Visual transition interval from pH 3.8 (yellow) to pH 5.4 (blue)
Clarity of solution Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR