Bromocresol green ACS

Supplier: Thermo Scientific Chemicals

Synonyms: 3,3-Bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

40293-0010 40293-0050 40293-0100
TS40293-0010EA 28 USD
TS40293-0010 TS40293-0050 TS40293-0100
Bromocresol green ACS
Bromocresol green
Formula: C₂₁H₁₄Br₄O₅S
Melting Pt: 225 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005874
CAS Number: 76-60-8
EINECS: 200-972-8
Merck Index: 13,01370
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12b4d4b3d992f8e86746537bf756af442752c853&vid=553ef1836a5b191a43558f8550e78cf2e424446e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Yellow to beige to light brown Powder
Infrared spectrum Conforms
Visual transition interval from pH 3.8 (yellow) to pH 5.4 (blue)
Clarity of solution Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR