Borane trimethylamine complex 97%

Supplier: Thermo Scientific Chemicals
Warning

40274-0250 40274-1000
TS40274-0250EA 107 USD
TS40274-0250 TS40274-1000
Borane trimethylamine complex 97%
Borane trimethylamine complex
Formula: C₃H₁₂BN
MW: 72.95 g/mol
Melting Pt: 92…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00012420
CAS Number: 75-22-9
EINECS: 200-850-4
UN: 1325
ADR: 4.1,II

Order Now

Specification Test Results

Appearance White Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 92 to 96 °C
Titration with Na2S2O3 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR