Betaine monohydrate 99+% for analysis

Supplier: Thermo Scientific Chemicals

40219-1000 40219-5000
TS40219-1000EA 72 USD
TS40219-1000 TS40219-5000
Betaine monohydrate 99+% for analysis
Betaine monohydrate
Formula: C₅H₁₁NO₂·1H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150010
CAS Number: 590-47-6
EINECS: 203-490-6

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying 11 % to 15 % (140°C, 3 h)
Sulfated ash ≤0.1 %
Solubility (10% in H2O) Clear colorless solution
Titration with HClO4 99.0 to 101.0 % (On dry substance)
Chloride (Cl) ≤200 ppm
Sulfate (SO4) ≤200 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR