Leuco malachite green, pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4,4'-Bis(dimethylamino)triphenylmethane, 4,4'-(Phenylmethylene)bis(N,N-dimethylaniline), 4,4'-Tetramethyldiaminotriphenylmethane, 4,4'-Benzylidenebis(N,N-dimethylaniline), Leucomalachite green

40211-0050 40211-0250
TS40211-0050EA 45 USD
TS40211-0050 TS40211-0250
Leuco malachite green, pure
Leuco malachite green
Formula: C₂₃H₂₆N₂
MW: 330.48 g/mol
Melting Pt: 100…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00008315
CAS Number: 129-73-7

Order Now

Specification Test Results

Appearance White to light green to light brown to light blue Powder
Infrared spectrum Conforms
UV spectrum Complies
TMS 0.03 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR