Acid orange 5 indicator grade

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: OrangeIV, Aniline yellow, 4-(4-Anilinophenylazo)benzenesulphonic acid sodium salt, CI 13080, Diphenylamine orange, Indicator, S. No.: 179, Orange IV, Tropaeolin OO

40126-0010 40126-0250 40126-1000
TS40126-0010EA 1649 USD
TS40126-0010 TS40126-0250 TS40126-1000
Acid orange 5 indicator grade
Acid orange 5
Formula: C₁₈H₁₄N₃NaO₃S
MW: 375.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038130
CAS Number: 554-73-4

Order Now

Specification Test Results

Appearance Ochre to orange Powder
Infrared spectrum Conforms
Visual transition interval from pH 1.4 (red) to pH 2.6 (yellow)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR