2-(Triphenylphosphoranylidene)propionaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Triphenylphosphoranylidene)propanal

39911-0010 39911-0100
TS39911-0010EA 38 USD
TS39911-0010 TS39911-0100
2-(Triphenylphosphoranylidene)propionaldehyde 98%
2-(Triphenylphosphoranylidene)propionaldehyde
Formula: C₂₁H₁₉OP
MW: 318.356 g/mol
Melting Pt: 213…217 °C
Storage Temperature: Ambient
MDL Number: MFCD00075596
CAS Number: 24720-64-7

Order Now

Specification Test Results

Appearance White to light beige Powder
Infrared spectrum Conforms
NMR (1H-NMR) ≥97.5 %
NMR (31P-NMR) ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR