3-Butynyl toluene-4-sulfonate 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Butynyl 4-methylbenzenesulphonate, 3-Butynyl p-toluenesulphonate

39078-0050 39078-0250
TS39078-0010EA 29.46 USD
TS39078-0010 TS39078-0050 TS39078-0250
3-Butynyl toluene-4-sulfonate 96%
3-Butynyl toluene-4-sulphonate
Formula: C₁₁H₁₂O₃S
MW: 224.28 g/mol
Density: 1.012 g/cm³
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041687
CAS Number: 23418-85-1

Order Now

Specification Test Results

Appearance Clear colorless to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5215 to 1.5255 (20°C, 589 nm)
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR