Allyl methyl carbonate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Carbonic acid Allyl Methyl Ester, Allyl Methyl Carbonate

38975-0050 38975-0250
TS38975-0050EA 87 USD
TS38975-0050 TS38975-0250
Allyl methyl carbonate 98%
Allyl methyl carbonate
Formula: C₅H₈O₃
MW: 116.1152 g/mol
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00134545
CAS Number: 35466-83-2
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to almost colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4050 to 1.4090 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR