[Hydroxy(tosyloxy)iodo]benzene 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Koser Reagent, Phenyliodosohydroxy Tosylate

38961-0050 38961-0250
TS38961-0050EA 43 USD
TS38961-0050 TS38961-0250
[Hydroxy(tosyloxy)iodo]benzene 97+%
[Hydroxy(tosyloxy)iodo]benzene
Formula: C₁₃H₁₃IO₄S
MW: 392.21 g/mol
Melting Pt: 131…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00011547
CAS Number: 27126-76-7

Order Now

Specification Test Results

Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR