Boron trifluoride dihydrate (65% BF₃)

Supplier: Thermo Scientific Chemicals
Danger

38871-1000 38871-5000
TS38871-1000EA 52 USD
TS38871-1000 TS38871-5000
Boron trifluoride dihydrate (65% BF₃)
Boron trifluoride dihydrate
Formula: BF₃·2H₂O
MW: 103.84 g/mol
Boiling Pt: 58…60 °C (1.5 mmHg)
Melting Pt: 6 °C
Density: 1.642 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149901
CAS Number: 13319-75-0
EINECS: 231-569-5
UN: 2851
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Assay 65.0 to 66.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR