trans-Bis(tributylstannyl)ethylene 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
trans-1,2-Bis(tri-n-butylstannyl)ethylene
38867-0010
38867-0050
TS38867-0010EA
96
USD
TS38867-0010
TS38867-0050
trans-Bis(tributylstannyl)ethylene 95%
trans-Bis(tributylstannyl)ethylene
Formula:
C₂₆H₅₆Sn₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD01631299 CAS Number: 14275-61-7 UN: 2788 ADR: 6.1,III |
Specification Test Results
Appearance | Colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5000 to 1.5030 (20°C, 589 nm) |
NMR | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...