Benzyltrimethylammonium tribromide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Mono(N,N,N-trimethyl-1-phenylmethanaminium) tribromide, Benzyl trimethyl ammonium tribromide

38832-0100 38832-0500
TS38832-0100EA 102 USD
TS38832-0100 TS38832-0500
Benzyltrimethylammonium tribromide 97%
Benzyltrimethylammonium tribromide
Formula: C₁₀H₁₆Br₃N
Melting Pt: 97…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00134423
CAS Number: 111865-47-5

Order Now

Specification Test Results

Appearance Yellow to orange Crystalline powder
Infrared spectrum Conforms
Titration Iodometric ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR