N-(2-Pyridyl)bis(trifluoromethanesulfonimide) 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N,N-Bis(trifluoromethylsulphonyl)-2-pyridylamine, 2-Pyridyltriflimide, 2-[N,N-Bis(trifluoromethylsulphonyl)amino]pyridine, 1,1,1-Trifluoro-N-(pyridin-2-yl)-N-((trifluoromethyl)sulphonyl)methanesulphonamide

38765-0050 38765-0250
TS38765-0050EA 122 USD
TS38765-0050 TS38765-0250
N-(2-Pyridyl)bis(trifluoromethanesulfonimide) 97%
N-(2-Pyridyl)bis(trifluoromethanesulfonimide)
Formula: C₇H₄F₆N₂O₄S₂
Boiling Pt: 97 °C (2 mmHg)
Melting Pt: 40…43 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191834
CAS Number: 145100-50-1
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow Powder or crystals
Infrared spectrum Conforms
Melting point 39 to 43 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR