1(2H)-Phthalazinone 98+%

Supplier: Thermo Scientific Chemicals
Warning

38457-0050 38457-0250 38457-1000 38457-5000
TS38457-0050EA 30 USD
TS38457-0050 TS38457-0250 TS38457-1000 TS38457-5000
1(2H)-Phthalazinone 98+%
1(2H)-Phthalazinone
Formula: C₈H₆N₂O
Melting Pt: 184…187 °C
Storage Temperature: Ambient
MDL Number: MFCD00006892
CAS Number: 119-39-1

Order Now

Specification Test Results

Appearance Off-white to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 184 to 187 °C
HPLC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR