Nitrosium hexafluorophosphate 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Nitrosonium hexafluorophosphate, Nitrosyl hexafluorophosphate

38373-0050 38373-0250
TS38373-0050EA 131 USD
TS38373-0050 TS38373-0250
Nitrosium hexafluorophosphate 95%
Nitrosium hexafluorophosphate
Formula: F₆NOP
MW: 174.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00040324
CAS Number: 16921-91-8
EINECS: 240-980-9
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to white Crystals
Assay ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR