Bis(dichlorophosphino)methane 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Phosphonous dichloride,P,P'-methylenebis-, Methylenebis(dichlorophosphine), dichloro(dichlorophosphanylmethyl)phosphane

38300-0010 38300-0050
TS38300-0010EA 77 USD
TS38300-0010 TS38300-0050
Bis(dichlorophosphino)methane 96%
Bis(dichlorophosphino)methane
Formula: CH₂Cl₄P₂
Boiling Pt: 101…105 °C (22 mmHg)
Density: 1.601 g/cm³
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD01073767
CAS Number: 28240-68-8
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Refractive index 1.5920 to 1.5950 (20°C, 589 nm)
Titration Argentometric ≥95 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR