Triphenylphosphine copper(I) hydride hexamer 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Stryker's reagent

38251-0010 38251-0050
TS38251-0010EA 92 USD
TS38251-0010 TS38251-0050
Triphenylphosphine copper(I) hydride hexamer 96%
Triphenylphosphine copper(I) hydride hexamer
Formula: C₁₀₈H₁₀₂Cu₆P₆
MW: 1967.08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00221518
CAS Number: 33636-93-0

Order Now

Specification Test Results

Appearance Red Powder or crystals
Elemental Analysis ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR